Assay Details
| Gene Symbol: CWF19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152434.2 | NP_689647.2 | 16 - 17 | 2467 | 95 | |
| XM_017017239.1 | XP_016872728.1 | 15 - 16 | 1785 | 95 | ||
| GenBank mRNA | AK056905.1 | - | 14 - 15 | 2119 | 95 | |
| AK126881.1 | - | 16 - 17 | 2458 | 95 | ||
| BC028043.2 | - | 3 - 4 | 316 | 95 | ||
| BC056241.1 | - | 4 - 5 | 562 | 95 | ||
| BC110439.1 | - | 14 - 15 | 2157 | 40001346 | 95 | |
| BC110440.1 | - | 13 - 14 | 2079 | 40001348 | 95 | |
| BC110441.1 | - | 13 - 14 | 2022 | 40001349 | 95 | |
| BC110442.1 | - | 13 - 14 | 2080 | 40001350 | 95 | |
| BC118669.1 | - | 13 - 14 | 2083 | 40012545 | 95 | |
| BX647832.1 | - | 16 - 17 | 2441 | 95 | ||
| BX648704.1 | - | 16 - 17 | 2446 | 95 | ||
| BX649022.1 | - | 16 - 17 | 2437 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002259.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11027009.hg.1 | ENST00000282251.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027009.hg.1 | ENST00000409771.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027009.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027009.hg.1 | TC11002259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | ENST00000282251.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | ENST00000431778.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | ENST00000532251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | NM_152434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193855.hg.1 | TC1100012208.hg.1 | Clariom D TC |

CWF19L2
Genomic Map