Assay Details
| Gene Symbol: INPP5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005539.4 | NP_005530.3 | 2 - 3 | 436 | 83 | |
| XM_017016205.1 | XP_016871694.1 | 2 - 3 | 431 | 83 | ||
| GenBank mRNA | AF273055.1 | - | 2 - 3 | 379 | 83 | |
| BC096280.2 | - | 3 - 4 | 180 | 40006844 | 83 | |
| BC096281.2 | - | 2 - 3 | 99 | 40006846 | 83 | |
| HY025461.1 | - | 2 - 3 | 437 | 83 | ||
| X77567.1 | - | 2 - 3 | 217 | 83 | ||
| Z31695.1 | - | 2 - 3 | 75 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000946.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC096281.2,Z31695.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011654.hg.1 | ENST00000368593.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011654.hg.1 | ENST00000368594.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011654.hg.1 | ENST00000368595.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011654.hg.1 | ENST00000416326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011654.hg.1 | ENST00000432898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011654.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011654.hg.1 | TC10000946.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164393.hg.1 | TC1000009414.hg.1 | Clariom D TC |

INPP5A
Genomic Map