Assay Details
| Gene Symbol: EXOSC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001998.2 | NP_001001998.1 | 19 - 20 | 2235 | 63 | |
| XM_005263475.2 | XP_005263532.2 | 19 - 20 | 2194 | 63 | ||
| XM_017001494.1 | XP_016856983.1 | 18 - 19 | 1920 | 63 | ||
| XM_017001495.1 | XP_016856984.1 | 18 - 19 | 1920 | 63 | ||
| XM_017001496.1 | XP_016856985.1 | 19 - 20 | 2265 | 63 | ||
| XM_017001497.1 | XP_016856986.1 | 19 - 20 | 2265 | 63 | ||
| GenBank mRNA | AB209236.1 | - | 15 - 16 | 3767 | 63 | |
| AK123365.1 | - | 7 - 8 | 1393 | 63 | ||
| AK296559.1 | - | 17 - 18 | 2735 | 63 | ||
| BC009908.2 | - | 15 - 16 | 1557 | 63 | ||
| BC039901.1 | - | 19 - 20 | 2176 | 5505500 | 63 | |
| BC073788.1 | - | 19 - 20 | 2211 | 5420763 | 63 | |
| CA307576.1 | - | 6 - 7 | 63 | |||
| X66113.1 | - | 19 - 20 | 2193 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

EXOSC10
Genomic Map