Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928881 | LOC101928881 | Anti-sense | Chr. 2 - 233012614 - 233015885 on NCBI Build GRCh38 | ||
| Downstream | neuraminidase 2 | NEU2 | SIAL2 | 9.8kb | Anti-sense | Chr. 2 - 233032672 - 233035057 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map