| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046219.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046219.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046219.hg.1 |
TC02002865.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046217.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046217.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046217.hg.1 |
TC02002865.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046218.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046218.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02046218.hg.1 |
TC02002865.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200217815.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217815.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217815.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217815.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217815.hg.1 |
TC0200016026.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016026.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217816.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217816.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217816.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217816.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217816.hg.1 |
TC0200016026.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200217813.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217813.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217813.hg.1 |
ENST00000305141.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200217813.hg.1 |
NM_006056.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200217813.hg.1 |
TC0200016026.hg.1 |
Clariom D TC |