Assay Details
| Gene Symbol: FAM129A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052966.3 | NP_443198.1 | 10 - 11 | 1564 | 65 | |
| XM_011509140.2 | XP_011507442.1 | 11 - 12 | 2439 | 65 | ||
| GenBank mRNA | AB050477.1 | - | 10 - 11 | 1490 | 65 | |
| AF288391.1 | - | 10 - 11 | 1528 | 65 | ||
| AK022527.1 | - | 10 - 11 | 1402 | 65 | ||
| AK095547.1 | - | 5 - 6 | 808 | 65 | ||
| AK311019.1 | - | 6 - 7 | 915 | 65 | ||
| AY550972.1 | - | 10 - 11 | 1403 | 65 | ||
| BC030531.1 | - | 10 - 11 | 1420 | 5212338 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003624.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01055746.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055746.hg.1 | ENST00000487074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055746.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055746.hg.1 | TC01003624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055747.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055747.hg.1 | ENST00000487074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055747.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055747.hg.1 | TC01003624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000417056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000461167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | ENST00000496299.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016678.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226529.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226529.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226529.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226529.hg.1 | ENST00000367511.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226529.hg.1 | ENST00000487074.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226529.hg.1 | NM_052966.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226529.hg.1 | TC0100016678.hg.1 | Clariom D TC |

FAM129A
Genomic Map