| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055763.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055763.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01055763.hg.1 |
TC01003624.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000496299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
ENST00000496299.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000417056.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000461167.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000487074.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000496299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
ENST00000496299.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016678.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226542.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100226542.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226542.hg.1 |
ENST00000367511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100226542.hg.1 |
NM_052966.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100226542.hg.1 |
TC0100016678.hg.1 |
Clariom D TC |