Assay Details
| Gene Symbol: COL9A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001852.3 | NP_001843.1 | 2 - 3 | 217 | 150 | |
| XM_006710365.3 | XP_006710428.1 | 3 - 4 | 325 | 150 | ||
| XM_017000332.1 | XP_016855821.1 | 2 - 3 | 259 | 150 | ||
| GenBank mRNA | BC136326.1 | - | 2 - 3 | 210 | 9020313 | 150 |
| BC136327.1 | - | 2 - 3 | 210 | 9020314 | 150 | |
More Information
Set Membership: |
|

COL9A2
Genomic Map