Assay Details
| Gene Symbol: RNF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007212.3 | NP_009143.1 | 1 - 2 | 229 | 149 | |
| XM_005245413.3 | XP_005245470.1 | 1 - 2 | 218 | 149 | ||
| GenBank mRNA | AF141327.1 | - | 1 - 2 | 221 | 149 | |
| AK091574.1 | - | 1 - 2 | 122 | 149 | ||
| BF183837.1 | - | 227 | 148 | |||
| BX338357.2 | - | 1 - 2 | 273 | 149 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001595.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF141327.1,AK091574.1,NM_007212.3,XM_005245413.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
gastrulation with mouth forming second
transcription, DNA-templated
germ cell development
anterior/posterior axis specification
protein sumoylation
spinal cord development
response to retinoic acid
histone H2A monoubiquitination
histone H2A-K119 monoubiquitination
negative regulation of sequence-specific DNA binding transcription factor activity
chromatin binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
RING-like zinc finger domain binding
negative regulation of transcription from RNA polymerase II promoter
mitotic cell cycle
gastrulation with mouth forming second
transcription, DNA-templated
germ cell development
anterior/posterior axis specification
protein sumoylation
spinal cord development
response to retinoic acid
histone H2A monoubiquitination
histone H2A-K119 monoubiquitination
negative regulation of sequence-specific DNA binding transcription factor activity
chromatin binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
RING-like zinc finger domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01024719.hg.1 | ENST00000367509.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024719.hg.1 | ENST00000367510.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024719.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024719.hg.1 | TC01001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024720.hg.1 | ENST00000367509.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024720.hg.1 | ENST00000367510.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024720.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024720.hg.1 | TC01001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024717.hg.1 | ENST00000367509.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024717.hg.1 | ENST00000367510.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024717.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024717.hg.1 | TC01001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009492.hg.1 | ENST00000442955.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024714.hg.1 | ENST00000367510.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024714.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024714.hg.1 | TC01001595.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100180143.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180143.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180143.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180143.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180143.hg.1 | TC0100010903.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010903.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180150.hg.1 | TC0100010903.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000453650.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180149.hg.1 | TC0100010903.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009492.hg.1 | ENST00000442955.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | ENST00000367509.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | ENST00000367510.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | ENST00000498201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | NM_007212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100180146.hg.1 | TC0100010903.hg.1 | Clariom D TC |

RNF2
Genomic Map