Assay Details
| Gene Symbol: CDH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252338.2 | NP_001239267.1 | 6 - 7 | 948 | 82 | |
| NM_001252339.2 | NP_001239268.1 | 6 - 7 | 1105 | 82 | ||
| NM_001794.4 | NP_001785.2 | 7 - 8 | 1133 | 82 | ||
| GenBank mRNA | AK091496.1 | - | 7 - 8 | 1329 | 82 | |
| AK124724.1 | - | 6 - 7 | 1088 | 82 | ||
| AK308345.1 | - | 6 - 7 | 947 | 82 | ||
| AK315192.1 | - | 7 - 8 | 1087 | 82 | ||
| BC101651.1 | - | 7 - 8 | 1133 | 8069157 | 82 | |
| BC112150.1 | - | 7 - 8 | 1133 | 8327618 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000481.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20006698.hg.1 | ENST00000360469.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006698.hg.1 | ENST00000536643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006698.hg.1 | ENST00000543233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006698.hg.1 | NM_001252338.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006698.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006698.hg.1 | NM_001794.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20006698.hg.1 | TC20000481.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | TC2000007973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000155481.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000543233.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000611855.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | ENST00000614565.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007973.hg.1 | NM_001252339.2 | Refseq |

CDH4
Genomic Map