| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005812.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005812.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01005812.hg.1 |
TC01000377.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007552.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152834.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152834.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152834.hg.1 |
ENST00000253063.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100152834.hg.1 |
NM_031459.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100152834.hg.1 |
TC0100007552.hg.1 |
Clariom D TC |