| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005011.hg.1 |
ENST00000333646.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005011.hg.1 |
ENST00000374080.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005011.hg.1 |
ENST00000374102.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005011.hg.1 |
ENST00000536756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005011.hg.1 |
NM_005120.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X005011.hg.1 |
TC0X000382.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
ENST00000333646.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
ENST00000374080.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
ENST00000374102.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
NM_005120.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
ENST00000333646.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
ENST00000374080.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
ENST00000374102.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
NM_005120.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151830.hg.1 |
TC0X00007557.hg.1 |
Clariom D TC |