| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019886.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019886.hg.1 |
ENST00000418360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019886.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019886.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019886.hg.1 |
TC01006333.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239701.hg.1 |
TC0100018303.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000368408.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000470294.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
ENST00000498667.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018303.hg.1 |
NM_004952.4 |
Refseq |