| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013506.hg.1 |
ENST00000329687.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013506.hg.1 |
ENST00000355443.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013506.hg.1 |
ENST00000507555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013506.hg.1 |
ENST00000514800.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013506.hg.1 |
NM_001120.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013506.hg.1 |
NM_001146069.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04013506.hg.1 |
TC04000978.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000329687.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000355443.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000503596.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000507272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000507555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000508276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000512781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000514031.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000514800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
NM_001120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
NM_001146069.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000329687.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000355443.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000503596.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000507272.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000507555.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000508276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000512781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000514031.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
ENST00000514800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
NM_001120.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
NM_001146069.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400167311.hg.1 |
TC0400009790.hg.1 |
Clariom D TC |