Assay Details
| Gene Symbol: CHGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275.3 | NP_001266.1 | 3 - 4 | 454 | 88 | |
| NM_001301690.1 | NP_001288619.1 | 3 - 4 | 454 | 88 | ||
| XM_011536370.1 | XP_011534672.1 | 4 - 5 | 806 | 88 | ||
| GenBank mRNA | AK223381.1 | - | 3 - 4 | 401 | 88 | |
| AK313757.1 | - | 3 - 4 | 405 | 88 | ||
| BC001059.2 | - | 3 - 4 | 254 | 3509603 | 88 | |
| BC006459.1 | - | 3 - 4 | 394 | 3503641 | 88 | |
| BT006869.1 | - | 3 - 4 | 194 | 88 | ||
| BX248263.1 | - | 3 - 4 | 378 | 88 | ||
| DA201149.1 | - | 3 - 4 | 454 | 88 | ||
| J03483.1 | - | 3 - 4 | 276 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000574.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
regulation of the force of heart contraction
mast cell chemotaxis
organelle organization
regulation of blood pressure
positive regulation of cAMP metabolic process
killing of cells of other organism
mast cell cytokine production
negative regulation of catecholamine secretion
ovulation cycle
mast cell degranulation
innate immune response
mast cell activation
negative regulation of vasodilation
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
defense response to fungus
positive regulation of cardiac muscle contraction
adrenergic receptor signaling pathway involved in cardiac muscle relaxation
positive regulation of phospholipase C-activating G-protein coupled receptor signaling pathway
negative regulation of neuron death
positive regulation of relaxation of cardiac muscle
positive regulation of dense core granule biogenesis
regulation of the force of heart contraction
mast cell chemotaxis
organelle organization
regulation of blood pressure
positive regulation of cAMP metabolic process
killing of cells of other organism
mast cell cytokine production
negative regulation of catecholamine secretion
ovulation cycle
mast cell degranulation
innate immune response
mast cell activation
negative regulation of vasodilation
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
defense response to fungus
positive regulation of cardiac muscle contraction
adrenergic receptor signaling pathway involved in cardiac muscle relaxation
positive regulation of phospholipase C-activating G-protein coupled receptor signaling pathway
negative regulation of neuron death
positive regulation of relaxation of cardiac muscle
positive regulation of dense core granule biogenesis
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | ENST00000556876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007010.hg.1 | ENST00000216492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007010.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007010.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007010.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007010.hg.1 | TC14000574.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | ENST00000556876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008021.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000216492.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000334654.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000553866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000556076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | ENST00000556098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | NM_001275.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | NM_001301690.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400156818.hg.1 | TC1400008021.hg.1 | Clariom D TC |

CHGA
Genomic Map