Assay Details
| Gene Symbol: LDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194436.2 | NP_919417.1 | 5 - 6 | 681 | 113 | |
| GenBank mRNA | AY092767.1 | - | 5 - 6 | 686 | 113 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001274.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017751.hg.1 | ENST00000300051.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017751.hg.1 | ENST00000450168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017751.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017751.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017751.hg.1 | TC16001274.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | TC1600010866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184513.hg.1 | NM_194436.2 | Refseq |

LDHD
Genomic Map