Assay Details
| Gene Symbol: LDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153486.3 | NP_705690.2 | 7 - 8 | 1073 | 92 | |
| NM_194436.2 | NP_919417.1 | 7 - 8 | 1004 | 92 | ||
| GenBank mRNA | BC040279.1 | - | 7 - 8 | 1038 | 5162826 | 92 |
| BC047902.1 | - | 7 - 8 | 1068 | 5762691 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001274.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16017749.hg.1 | ENST00000300051.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017749.hg.1 | ENST00000450168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017749.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017749.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16017749.hg.1 | TC16001274.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | TC1600010866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600184511.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000300051.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000450168.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000568164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | ENST00000569876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_153486.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010866.hg.1 | NM_194436.2 | Refseq |

LDHD
Genomic Map