Assay Details
| Gene Symbol: MEAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022756.5 | NP_073593.2 | 6 - 7 | 640 | 79 | |
| NR_073092.1 | 7 - 8 | 729 | 79 | |||
| GenBank mRNA | AK308118.1 | - | 6 - 7 | 574 | 79 | |
| AK309663.1 | - | 7 - 8 | 690 | 79 | ||
| BX538212.1 | - | 6 - 7 | 576 | 79 | ||
| BX640719.1 | - | 6 - 7 | 585 | 79 | ||
More Information
Set Membership: |
|

MEAF6
Genomic Map