Assay Details
| Gene Symbol: IP6K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142883.1 | NP_001136355.1 | 2 - 3 | 359 | 65 | |
| GenBank mRNA | AK289935.1 | - | 2 - 3 | 279 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001592.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK289935.1,NM_001142883.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
phosphatidylinositol metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 6-kinase activity
inositol hexakisphosphate 5-kinase activity
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
protein phosphorylation
inositol phosphate biosynthetic process
inositol phosphate metabolic process
phosphatidylinositol metabolic process
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 6-kinase activity
inositol hexakisphosphate 5-kinase activity
ATP binding
inositol-1,4,5-trisphosphate 3-kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06019367.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06019367.hg.1 | ENST00000451316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06019367.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06019367.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06019367.hg.1 | TC06001592.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179232.hg.1 | TC0600011561.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600179233.hg.1 | TC0600011561.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000293756.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | ENST00000451316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_001142883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011561.hg.1 | NM_054111.4 | Refseq |

IP6K3
Genomic Map