Assay Details
| Gene Symbol: PALM2-AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ312217.1 | - | 7 - 8 | 1237 | 70 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000302798.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000374530.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | ENST00000413420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_007203.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012158.hg.1 | NM_147150.2 | Refseq |

PALM2-AKAP2
Genomic Map