Assay Details
| Gene Symbol: PALM2-AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007203.4 | NP_009134.1 | 7 - 8 | 767 | 77 | |
| NM_147150.2 | NP_671492.1 | 7 - 8 | 767 | 77 | ||
| GenBank mRNA | AB023137.1 | - | 7 - 8 | 767 | 77 | |
| AJ303079.1 | - | 7 - 8 | 755 | 77 | ||
| BC140818.1 | - | 7 - 8 | 608 | 9021686 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

PALM2-AKAP2
Genomic Map