Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FGF13 antisense RNA 1 | FGF13-AS1 | -191.8kb | Anti-sense | Chr. X - 138712107 - 138716605 on NCBI Build GRCh38 | |
| Downstream | steroid 5 alpha-reductase 1 pseudogene 1 | SRD5A1P1 | SRD5AP1 | 537.1kb | Anti-sense | Chr. X - 139446819 - 139448973 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map