Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146051.1 | NP_001139523.1 | 5 - 6 | 683 | 63 | |
| NM_031463.4 | NP_113651.4 | 4 - 5 | 848 | 63 | ||
| XM_005256189.2 | XP_005256246.1 | 4 - 5 | 816 | 63 | ||
| GenBank mRNA | AF237684.1 | - | 3 - 4 | 712 | 63 | |
| AK074878.1 | - | 4 - 5 | 812 | 63 | ||
| AK299798.1 | - | 5 - 6 | 657 | 63 | ||
| AY028377.1 | - | 3 - 4 | 712 | 63 | ||
| BC018084.1 | - | 4 - 5 | 841 | 4814348 | 63 | |
| BC104218.1 | - | 3 - 4 | 729 | 40032007 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001308.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018135.hg.1 | ENST00000219439.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018135.hg.1 | ENST00000434463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018135.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018135.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018135.hg.1 | TC16001308.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | TC1600011047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185813.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | TC1600011047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185811.hg.1 | NM_031463.4 | Refseq |

HSDL1
Genomic Map