Assay Details
| Gene Symbol: INTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324475.1 | NP_001311404.1 | 1 - 2 | 152 | 142 | |
| GenBank mRNA | AK290025.1 | - | 1 - 2 | 146 | 142 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK290025.1, NM_001324475.1 transcripts.
Set Membership: |
|

INTS3
Genomic Map