Assay Details
| Gene Symbol: UBE3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014671.2 | NP_055486.2 | 2 - 3 | 436 | 137 | |
| GenBank mRNA | AK295138.1 | - | 2 - 3 | 116 | 137 | |
| BC014029.2 | - | 3 - 4 | 196 | 4541634 | 137 | |
| D13635.1 | - | 2 - 3 | 427 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK295138.1 transcripts.
Set Membership: |
|

UBE3C
Genomic Map