Assay Details
| Gene Symbol: PAQR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104554.1 | NP_001098024.1 | 1 - 2 | 391 | 85 | |
| GenBank mRNA | AY424283.1 | - | 1 - 2 | 391 | 85 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000635.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY424283.1,NM_001104554.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000384665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | ENST00000561250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15005376.hg.1 | ENST00000340965.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15005376.hg.1 | ENST00000561153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15005376.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15005376.hg.1 | TC15000635.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500154663.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154663.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500154663.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154663.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500154663.hg.1 | TC1500007695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000340965.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000384665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000395407.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000558684.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | ENST00000561250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_001104554.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007695.hg.1 | NM_017705.3 | Refseq |

PAQR5
Genomic Map