| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000286355.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000377928.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000522949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000522949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000522949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000522949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000522949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
ENST00000522949.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011844.hg.1 |
NM_001115.2 |
Refseq |