Assay Details
| Gene Symbol: CABLES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031215.2 | NP_112492.2 | 9 - 10 | 90 | ||
| GenBank mRNA | BC003122.1 | - | 7 - 8 | 836 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001009.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20013853.hg.1 | ENST00000279101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013853.hg.1 | ENST00000370560.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013853.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013853.hg.1 | TC20001009.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013854.hg.1 | ENST00000279101.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013854.hg.1 | ENST00000370560.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013854.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013854.hg.1 | TC20001009.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000167998.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167998.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167998.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167998.hg.1 | TC2000009748.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000167998.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167998.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167998.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167997.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167997.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167997.hg.1 | TC2000009748.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000167997.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167997.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167996.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167996.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000167996.hg.1 | TC2000009748.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000167996.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000167996.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000279101.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | ENST00000453274.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009748.hg.1 | NM_031215.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 213740 | - | - |

CABLES2
Genomic Map