Assay Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122962.1 | NP_001116434.1 | 4 - 5 | 1086 | 79 | |
| NM_001134836.1 | NP_001128308.1 | 4 - 5 | 792 | 79 | ||
| GenBank mRNA | AK095499.1 | - | 4 - 5 | 895 | 79 | |
| AK297072.1 | - | 4 - 5 | 686 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20001743.hg.1
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000359801.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000381630.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000444444.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000481731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000486775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608073.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000608747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | ENST00000609796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001122962.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009968.hg.1 | NM_001134836.1 | Refseq |

SIRPB2
Genomic Map