Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
ENST00000261678.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
ENST00000344113.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
ENST00000357395.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
ENST00000358025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
ENST00000554584.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
NM_015180.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
NM_182914.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004280.hg.1 |
TC14000386.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000344113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000357395.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000358025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000554584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000557005.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
NM_015180.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
NM_182914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000344113.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000357395.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000358025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000554584.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
ENST00000557005.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
NM_015180.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
NM_182914.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400151734.hg.1 |
TC1400007430.hg.1 |
Clariom D TC |