Assay Details
| Gene Symbol: NHSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011087.1 | XP_016866576.1 | 1 | 181 | 158 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011087.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000342260.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000343505.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000426841.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000427025.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000468095.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000479393.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000533765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | ENST00000534376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_001144060.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014348.hg.1 | NM_020464.1 | Refseq |

NHSL1
Genomic Map