Assay Details
| Gene Symbol: CNKSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297647.1 | NP_001284576.1 | 8 - 9 | 830 | 91 | |
| XM_011540485.2 | XP_011538787.1 | 5 - 6 | 319 | 91 | ||
| GenBank mRNA | BC011604.2 | - | 8 - 9 | 811 | 3139227 | 91 |
| BC012797.2 | - | 8 - 9 | 811 | 3505293 | 91 | |
| BT006900.1 | - | 8 - 9 | 772 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011540485.2 transcripts.
Set Membership: |
|

CNKSR1
Genomic Map