Assay Details
| Gene Symbol: FAM19A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040286.1 | - | 1 - 2 | 83 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC040286.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000416284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000500610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000500610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000500610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000500610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000500610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547919.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000547941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000548541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000548541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000548541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000548780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000548780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000548780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000548780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549379.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000549958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550003.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000550396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000551619.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000552075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000552075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000552075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000552075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000552075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | ENST00000552075.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011037.hg.1 | NM_178539.4 | Refseq |

FAM19A2
Genomic Map