Assay Details
| Gene Symbol: CDH20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031891.3 | NP_114097.2 | 5 - 6 | 1227 | 107 | |
| GenBank mRNA | AF217289.1 | - | 4 - 5 | 841 | 107 | |
| AK290190.1 | - | 5 - 6 | 1054 | 107 | ||
| BC101047.1 | - | 4 - 5 | 841 | 40007940 | 107 | |
| BC101048.1 | - | 4 - 5 | 841 | 40007941 | 107 | |
| BC101049.1 | - | 4 - 5 | 841 | 40007942 | 107 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000217.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000587582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000587582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | ENST00000587582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002468.hg.1 | ENST00000262717.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002468.hg.1 | ENST00000536675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002468.hg.1 | ENST00000538374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002468.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002468.hg.1 | TC18000217.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000587582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000587582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | ENST00000587582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007490.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | NM_031891.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | TC1800007490.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | ENST00000262717.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | ENST00000536675.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | ENST00000538374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151348.hg.1 | NM_031891.3 | Refseq |

CDH20
Genomic Map