Assay Details
| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000855.2 | NP_000846.1 | 7 - 8 | 2376 | 74 | |
| NM_001256424.1 | NP_001243353.1 | 8 - 9 | 2469 | 74 | ||
| GenBank mRNA | BC130484.1 | - | 7 - 8 | 2082 | 40146515 | 74 |
| BC130488.1 | - | 7 - 8 | 2082 | 40146519 | 74 | |
| BC144033.1 | - | 8 - 9 | 2145 | 9052553 | 74 | |
| X63282.1 | - | 7 - 8 | 2376 | 74 | ||
| Z50053.1 | - | 8 - 9 | 2469 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002258.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026991.hg.1 | ENST00000282249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026991.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026991.hg.1 | ENST00000526355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026991.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026991.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026991.hg.1 | TC11002258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026990.hg.1 | ENST00000282249.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026990.hg.1 | ENST00000526355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026990.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026990.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026990.hg.1 | TC11002258.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026989.hg.1 | ENST00000526355.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026989.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026989.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11026989.hg.1 | TC11002258.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193803.hg.1 | TC1100012197.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193804.hg.1 | TC1100012197.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193802.hg.1 | TC1100012197.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000282249.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000347596.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012197.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193801.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193801.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193801.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193801.hg.1 | ENST00000526355.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100193801.hg.1 | NM_000855.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193801.hg.1 | NM_001256424.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100193801.hg.1 | TC1100012197.hg.1 | Clariom D TC |

GUCY1A2
Genomic Map