| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050246.hg.1 |
ENST00000359215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050246.hg.1 |
ENST00000368718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050246.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050246.hg.1 |
TC01003265.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050247.hg.1 |
ENST00000359215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050247.hg.1 |
ENST00000368718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050247.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050247.hg.1 |
TC01003265.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050245.hg.1 |
ENST00000359215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050245.hg.1 |
ENST00000368718.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050245.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050245.hg.1 |
TC01003265.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368717.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015865.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219159.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219159.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219159.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219159.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219159.hg.1 |
TC0100015865.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219161.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219161.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219161.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219161.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219161.hg.1 |
TC0100015865.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100219160.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219160.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219160.hg.1 |
ENST00000368718.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100219160.hg.1 |
NM_002962.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100219160.hg.1 |
TC0100015865.hg.1 |
Clariom D TC |