Assay Details
| Gene Symbol: SCGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006998.3 | NP_008929.2 | 6 - 7 | 675 | 110 | |
| XM_017010181.1 | XP_016865670.1 | 5 - 6 | 387 | 110 | ||
| GenBank mRNA | AK289477.1 | - | 6 - 7 | 730 | 110 | |
| BC000336.2 | - | 6 - 7 | 642 | 2822613 | 110 | |
| BC003036.1 | - | 6 - 7 | 642 | 2822613 | 110 | |
| Y16752.1 | - | 6 - 7 | 628 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000157.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06001600.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06001600.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06001600.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06001600.hg.1 | TC06000157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06001601.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06001601.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06001601.hg.1 | TC06000157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149165.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149165.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149165.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149165.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149165.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149165.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149165.hg.1 | TC0600007244.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007244.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | ENST00000334979.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | ENST00000377961.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | ENST00000612225.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | NM_006998.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600149164.hg.1 | TC0600007244.hg.1 | Clariom D TC |

SCGN
Genomic Map