Assay Details
| Gene Symbol: PLA2G12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF332892.1 | - | 2 - 3 | 437 | 66 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000243501.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000502772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | ENST00000507961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011579.hg.1 | NM_030821.4 | Refseq |

PLA2G12A
Genomic Map