| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014415.hg.1 |
ENST00000296048.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014415.hg.1 |
ENST00000345003.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014415.hg.1 |
ENST00000483267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014415.hg.1 |
NM_001184720.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014415.hg.1 |
NM_004130.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03014415.hg.1 |
TC03000801.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000296048.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000345003.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000461191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000469873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000483267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000627418.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
NM_001184720.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
NM_004130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000296048.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000345003.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000461191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000469873.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000483267.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
ENST00000627418.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
NM_001184720.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
NM_004130.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300165440.hg.1 |
TC0300009126.hg.1 |
Clariom D TC |