Assay Details
| Gene Symbol: DHTKD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018706.6 | NP_061176.3 | 11 - 12 | 2166 | 87 | |
| GenBank mRNA | AK309810.1 | - | 11 - 12 | 2074 | 87 | |
| BC002477.2 | - | 11 - 12 | 2111 | 3347913 | 87 | |
| BC007955.2 | - | 11 - 12 | 2093 | 4302613 | 87 | |
| BU855972.1 | - | 3 - 4 | 377 | 87 | ||
| CR749726.1 | - | 6 - 7 | 914 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000089.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000936.hg.1 | ENST00000263035.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000936.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10000936.hg.1 | TC10000089.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146381.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146381.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146381.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146381.hg.1 | ENST00000263035.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146381.hg.1 | ENST00000448829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146381.hg.1 | NM_018706.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146381.hg.1 | TC1000006794.hg.1 | Clariom D TC |

DHTKD1
Genomic Map