Assay Details
| Gene Symbol: TRPV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019841.6 | NP_062815.3 | 1 - 2 | 389 | 62 | |
| GenBank mRNA | AF304464.1 | - | 1 - 2 | 474 | 62 | |
| AJ271207.2 | - | 1 - 2 | 125 | 62 | ||
| AJ487965.2 | - | 1 - 2 | 127 | 62 | ||
| BC034740.1 | - | 1 - 2 | 389 | 5186668 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001964.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030275.hg.1 | ENST00000265310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030275.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030275.hg.1 | NM_019841.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030275.hg.1 | TC07001964.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030273.hg.1 | ENST00000265310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030273.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030273.hg.1 | NM_019841.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030273.hg.1 | TC07001964.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030274.hg.1 | ENST00000265310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030274.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030274.hg.1 | NM_019841.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07030274.hg.1 | TC07001964.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700192860.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192860.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192860.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192860.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192860.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192860.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192860.hg.1 | TC0700012869.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700192862.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192862.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192862.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192862.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192862.hg.1 | TC0700012869.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012869.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192859.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192859.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192859.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192859.hg.1 | ENST00000265310.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192859.hg.1 | ENST00000439304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192859.hg.1 | ENST00000442623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192859.hg.1 | TC0700012869.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 138425 | - | - |

TRPV5
Genomic Map