Assay Details
| Gene Symbol: WDR83OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016145.3 | NP_057229.1 | 1 - 2 | 303 | 96 | |
| XM_017026864.1 | XP_016882353.1 | 2 - 3 | 1297 | 96 | ||
| GenBank mRNA | AF059620.1 | - | 1 - 2 | 71 | 96 | |
| AF078861.1 | - | 1 - 2 | 288 | 96 | ||
| AF151898.1 | - | 1 - 2 | 287 | 96 | ||
| AK091030.1 | - | 1 - 2 | 2008 | 96 | ||
| AK311944.1 | - | 1 - 2 | 67 | 96 | ||
| BC001192.1 | - | 1 - 2 | 51 | 96 | ||
| BC042919.1 | - | 1 - 2 | 72 | 5932109 | 96 | |
| BC061912.1 | - | 1 - 2 | 72 | 4444565 | 96 | |
More Information
Set Membership: |
|

WDR83OS
Genomic Map