Assay Details
| Gene Symbol: WDR83OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016145.3 | NP_057229.1 | 2 - 3 | 408 | 69 | |
| XM_017026864.1 | XP_016882353.1 | 3 - 4 | 1402 | 69 | ||
| GenBank mRNA | AF059620.1 | - | 2 - 3 | 176 | 69 | |
| AF078861.1 | - | 2 - 3 | 393 | 69 | ||
| AF151898.1 | - | 2 - 3 | 392 | 69 | ||
| AK091030.1 | - | 2 - 3 | 2113 | 69 | ||
| AK311944.1 | - | 2 - 3 | 172 | 69 | ||
| BC001192.1 | - | 2 - 3 | 156 | 69 | ||
| BC042919.1 | - | 2 - 3 | 177 | 5932109 | 69 | |
| BC061912.1 | - | 2 - 3 | 177 | 4444565 | 69 | |
More Information
Set Membership: |
|

WDR83OS
Genomic Map