Assay Details
| Gene Symbol: TMEM131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015348.1 | NP_056163.1 | 25 - 26 | 2948 | 106 | |
| XM_005263909.1 | XP_005263966.1 | 26 - 27 | 3034 | 106 | ||
| XM_005263910.1 | XP_005263967.1 | 26 - 27 | 3034 | 106 | ||
| XM_005263911.1 | XP_005263968.1 | 25 - 26 | 2917 | 106 | ||
| XM_005263912.3 | XP_005263969.1 | 24 - 25 | 3122 | 106 | ||
| XM_011510869.1 | XP_011509171.1 | 26 - 27 | 3034 | 106 | ||
| GenBank mRNA | D87446.1 | - | 24 - 25 | 2486 | 106 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002123.hg.1
Set Membership: |
|

TMEM131
Genomic Map