Assay Details
| Gene Symbol: TMEM131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015348.1 | NP_056163.1 | 34 - 35 | 4799 | 73 | |
| XM_005263909.1 | XP_005263966.1 | 35 - 36 | 4885 | 73 | ||
| XM_005263910.1 | XP_005263967.1 | 35 - 36 | 4885 | 73 | ||
| XM_005263911.1 | XP_005263968.1 | 34 - 35 | 4768 | 73 | ||
| XM_005263912.3 | XP_005263969.1 | 33 - 34 | 4973 | 73 | ||
| XM_011510869.1 | XP_011509171.1 | 34 - 35 | 4699 | 73 | ||
| GenBank mRNA | AK025852.1 | - | 4 - 5 | 447 | 73 | |
| AK098792.1 | - | 3 - 4 | 622 | 73 | ||
| D87446.1 | - | 33 - 34 | 4337 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002123.hg.1
Set Membership: |
|

TMEM131
Genomic Map