Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929357 | HP09053 | -109.3kb | Anti-sense | Chr. 3 - 99816632 - 99823865 on NCBI Build GRCh38 | |
| Downstream | microRNA 3921 | MIR3921 | 29.0kb | Anti-sense | Chr. 3 - 99964314 - 99964398 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map