Assay Details
| Gene Symbol: ATF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028807.1 | XP_016884296.1 | 1 - 2 | 96 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017028807.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
gluconeogenesis
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular amino acid metabolic process
gamma-aminobutyric acid signaling pathway
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
negative regulation of translational initiation in response to stress
circadian regulation of gene expression
cellular response to amino acid starvation
cellular response to UV
response to endoplasmic reticulum stress
positive regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of transcription from RNA polymerase II promoter in response to oxidative stress
PERK-mediated unfolded protein response
cellular response to glucose starvation
mRNA transcription from RNA polymerase II promoter
positive regulation of apoptotic process
negative regulation of potassium ion transport
positive regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase I promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter in response to arsenic-containing substance
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
negative regulation of oxidative stress-induced neuron death
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
response to manganese-induced endoplasmic reticulum stress
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
leucine zipper domain binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
gluconeogenesis
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular amino acid metabolic process
gamma-aminobutyric acid signaling pathway
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
negative regulation of translational initiation in response to stress
circadian regulation of gene expression
cellular response to amino acid starvation
cellular response to UV
response to endoplasmic reticulum stress
positive regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of transcription from RNA polymerase II promoter in response to oxidative stress
PERK-mediated unfolded protein response
cellular response to glucose starvation
mRNA transcription from RNA polymerase II promoter
positive regulation of apoptotic process
negative regulation of potassium ion transport
positive regulation of neuron apoptotic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase I promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter in response to arsenic-containing substance
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
negative regulation of oxidative stress-induced neuron death
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
response to manganese-induced endoplasmic reticulum stress
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
leucine zipper domain binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000337304.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000396680.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | ENST00000404241.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_001675.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_182810.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007406.hg.1 | NM_182810.2 | Refseq |

ATF4
Genomic Map