| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14011519.hg.1 |
ENST00000261312.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14011519.hg.1 |
ENST00000399332.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14011519.hg.1 |
ENST00000553700.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14011519.hg.1 |
NM_015382.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14011519.hg.1 |
TC14001009.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000399332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000553700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000554882.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000555311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000555843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000611816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
NM_015382.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000399332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000553700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000554882.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000555311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000555843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
ENST00000611816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
NM_015382.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400165073.hg.1 |
TC1400008844.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000399332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000553700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000554882.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000555311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000555843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000611816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
NM_015382.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000399332.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000553700.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000554882.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000555311.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000555843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
ENST00000611816.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
NM_015382.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400165072.hg.1 |
TC1400008844.hg.1 |
Clariom D TC |