| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013558.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013558.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22013558.hg.1 |
TC22000798.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008717.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200163264.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163264.hg.1 |
NM_014293.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200163264.hg.1 |
TC2200008717.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200163264.hg.1 |
ENST00000333039.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200163264.hg.1 |
NM_014293.3 |
Refseq |