Assay Details
| Gene Symbol: FAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028034.3 | 2 - 3 | 510 | 73 | ||
| NR_028035.3 | 2 - 3 | 510 | 73 | |||
| NR_135315.1 | 2 - 3 | 501 | 73 | |||
| GenBank mRNA | AV715411.1 | - | 2 - 3 | 346 | 73 | |
| X83490.1 | - | 2 - 3 | 217 | 73 | ||
| X83491.1 | - | 2 - 3 | 217 | 73 | ||
| Z47994.1 | - | 2 - 3 | 192 | 73 | ||
| Z47995.1 | - | 2 - 3 | 192 | 73 | ||
More Information
Set Membership: |
|

FAS
Genomic Map